Merck
  • Study of artemisinin and sugar accumulation in Artemisia vulgaris and Artemisia dracunculus "hairy" root cultures.
Study of artemisinin and sugar accumulation in Artemisia vulgaris and Artemisia dracunculus "hairy" root cultures.
Preparative biochemistry & biotechnology (2017-06-24)
Kateryna O Drobot, Nadiia A Matvieieva, Andriy M Ostapchuk, Maxim A Kharkhota, Volodymyr P Duplij
ABSTRACT
We studied the effect of genetic transformation on biologically active compound (artemisinin and its co-products (ART) as well as sugars) accumulation in Artemisia vulgaris and Artemisia dracunculus "hairy" root cultures. Glucose, fructose, sucrose, and mannitol were accumulated in A. vulgaris and A. dracunculus "hairy" root lines. Genetic transformation has led in some cases to the sugar content increasing or appearing of nonrelevant for the control plant carbohydrates. Sucrose content was 1.6 times higher in A. vulgaris "hairy" root lines. Fructose content was found to be 3.4 times higher in A. dracunculus "hairy" root cultures than in the control roots. The accumulation of mannitol was a special feature of the leaves of A. vulgaris and A. dracunculus control roots. A. vulgaris "hairy" root lines differed also in ART accumulation level. The increase of ART content up to 1.02 mg/g DW in comparison with the nontransformed roots (up to 0.687 mg/g DW) was observed. Thus, Agrobacterium rhizogenes-mediated genetic transformation can be used for obtaining of A. vulgaris and A. dracunculus "hairy" root culture produced ART and sugars in a higher amount than mother plants.