Merck
  • Spectroscopic Physicochemical and Photophysical Investigation of Biologically Active 2-oxo-quinoline-3-carbonitrile Derivative.
Spectroscopic Physicochemical and Photophysical Investigation of Biologically Active 2-oxo-quinoline-3-carbonitrile Derivative.
Journal of fluorescence (2017-01-23)
Mohie E M Zayed, Parveen Kumar
ABSTRACT
4-(2,3,4-trimethoxyphenyl)-8-methoxy-2-oxo-1,2,5,6 tetrahydrobenzo [h] quinoline-3-carbonitrile (TMTQ) dye was synthesized by one-pot multicomponent reactions (MCRs) of 2,3,4 trimethoxybenzaldehyd, ethyl cyanoacetate, 6-methoxy-1,2,3,4-tetrahydro-naphthalin-1-one and ammonium acetate under microwave irradiation. The structures of the synthesized compound was established by spectroscopic (FT-IR,
Sigma-Aldrich
Ethyl cyanoacetate, ≥98%
Pricing and availability is not currently available.