Merck
Transcription Factor RUNX1 Regulates Platelet
Circulation (2017-07-06)
Guangfen Mao, Natthapol Songdej, Deepak Voora, Lawrence E Goldfinger, Fabiola E Del Carpio-Cano, Rachel A Myers, A Koneti Rao
ABSTRACT
PCTP (phosphatidylcholine transfer protein) regulates the intermembrane transfer of phosphatidylcholine. Higher platelet PCTP expression is associated with increased platelet responses on activation of protease-activated receptor 4 thrombin receptors noted in black subjects compared with white subjects. Little is known about the regulation of platelet We pursued the hypothesis that Platelet PCTP protein in the patient was reduced by ≈50%. DNA-protein binding studies showed RUNX1 binding to consensus sites in ≈1 kB of
Sigma-Aldrich
MISSION® esiRNA, targeting human RUNX1
Sigma-Aldrich
MISSION® esiRNA, targeting human PCTP