Merck
  • Screening Natural Content of Water-Soluble B Vitamins in Fish: Enzymatic Extraction, HILIC Separation, and Tandem Mass Spectrometric Determination.
Screening Natural Content of Water-Soluble B Vitamins in Fish: Enzymatic Extraction, HILIC Separation, and Tandem Mass Spectrometric Determination.
Journal of AOAC International (2017-03-18)
Niladri Sekhar Chatterjee, K Ashok Kumar, K K Ajeeshkumar, K R Remya Kumari, K V Vishnu, Rangasamy Anandan, Suseela Mathew, C N Ravishankar
ABSTRACT
Despite the potential of LC with tandem MS (MS/MS) in improving sensitivity and selectivity, analytical methods are scarce for the determination of protein-bound and phosphorylated forms of B vitamins in food. This prompted us to develop a method for LC-MS/MS determination of naturally occurring nicotinamide, nicotinic acid, thiamine, pyridoxine, riboflavin, pantothenic acid, biotin, folic acid, and cyanocobalamin in fish. Baseline separation of the vitamins was achieved in a hydrophilic interaction LC condition. An ultrasonication-assisted enzymatic extraction protocol for sample preparation was optimized and validated. The time required for extraction was significantly reduced (to 4 h), while maintaining good extraction efficiency. Acetonitrile content (80%, v/v) in the prepared sample was found to be optimum for excellent peak shape and sensitivity. The dynamic linear range of the vitamins ranged from 2.5 to 500 ng/g, and the regression coefficient values were greater than 0.99. LOQ values ranged from 0.4 to 50 ng/g for the different vitamins. The spike recovery values at 50 and 100 ng/g ranged from 87.5 to 97.5%. The intra- and interday precision values were satisfactory. Accuracy of the developed method was determined by analysis of a Certified Reference Material. The method could also be used for unambiguous determination of the natural content of the target vitamins in fish.
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Pricing and availability is not currently available.