Merck
  • Regulated in Development and DNA Damage Response 1 Deficiency Impairs Autophagy and Mitochondrial Biogenesis in Articular Cartilage and Increases the Severity of Experimental Osteoarthritis.
Regulated in Development and DNA Damage Response 1 Deficiency Impairs Autophagy and Mitochondrial Biogenesis in Articular Cartilage and Increases the Severity of Experimental Osteoarthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2017-03-24)
Oscar Alvarez-Garcia, Tokio Matsuzaki, Merissa Olmer, Lars Plate, Jeffery W Kelly, Martin K Lotz
ABSTRACT
Regulated in development and DNA damage response 1 (REDD1) is an endogenous inhibitor of mechanistic target of rapamycin (mTOR) that regulates cellular stress responses. REDD1 expression is decreased in aged and osteoarthritic (OA) cartilage, and it regulates mTOR signaling and autophagy in articular chondrocytes in vitro. This study was undertaken to investigate the effects of REDD1 deletion in vivo using a mouse model of experimental OA. OA severity was histologically assessed in 4-month-old wild-type and REDD1 REDD1 deficiency increased the severity of changes in cartilage, menisci, subchondral bone, and synovium in the DMM model of OA. Chondrocyte death was increased in the cartilage of REDD1 Our findings suggest that REDD1 is a key mediator of cartilage homeostasis through regulation of autophagy and mitochondrial biogenesis and that REDD1 deficiency exacerbates the severity of injury-induced OA.
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human DDIT4