Merck
  • Elevated FOXG1 and SOX2 in glioblastoma enforces neural stem cell identity through transcriptional control of cell cycle and epigenetic regulators.
Elevated FOXG1 and SOX2 in glioblastoma enforces neural stem cell identity through transcriptional control of cell cycle and epigenetic regulators.
Genes & development (2017-05-04)
Harry Bulstrode, Ewan Johnstone, Maria Angeles Marques-Torrejon, Kirsty M Ferguson, Raul Bardini Bressan, Carla Blin, Vivien Grant, Sabine Gogolok, Ester Gangoso, Sladjana Gagrica, Christine Ender, Vassiliki Fotaki, Duncan Sproul, Paul Bertone, Steven M Pollard
ABSTRACT
Glioblastoma multiforme (GBM) is an aggressive brain tumor driven by cells with hallmarks of neural stem (NS) cells. GBM stem cells frequently express high levels of the transcription factors FOXG1 and SOX2. Here we show that increased expression of these factors restricts astrocyte differentiation and can trigger dedifferentiation to a proliferative NS cell state. Transcriptional targets include cell cycle and epigenetic regulators (e.g.,
Sigma-Aldrich
NSC 632839, ≥98% (HPLC)