Merck
  • Actin turnover maintains actin filament homeostasis during cytokinetic ring contraction.
Actin turnover maintains actin filament homeostasis during cytokinetic ring contraction.
The Journal of cell biology (2017-06-29)
Ting Gang Chew, Junqi Huang, Saravanan Palani, Ruth Sommese, Anton Kamnev, Tomoyuki Hatano, Ying Gu, Snezhana Oliferenko, Sivaraj Sivaramakrishnan, Mohan K Balasubramanian
ABSTRACT
Cytokinesis in many eukaryotes involves a tension-generating actomyosin-based contractile ring. Many components of actomyosin rings turn over during contraction, although the significance of this turnover has remained enigmatic. Here, using
Sigma-Aldrich
Adenosine 5′-triphosphate disodium salt solution, Crystalline ATP, HPLC purified, aqueous solution for RNA transcription
Sigma-Aldrich
Adenosine 5′-(β,γ-imido)triphosphate lithium salt hydrate, ≥93% (HPLC), powder
Sigma-Aldrich
CK-666, ≥98% (HPLC), powder
Sigma-Aldrich
Escort IV Transfection Reagent, Lipid reagent for transient and stable transfection of mammalian and insect cells.