Merck
  • Quantitative methylene blue decolourisation assays as rapid screening tools for assessing the efficiency of catalytic reactions.
Quantitative methylene blue decolourisation assays as rapid screening tools for assessing the efficiency of catalytic reactions.
Chemosphere (2017-02-23)
Jan Kruid, Ronen Fogel, Janice Leigh Limson
ABSTRACT
Identifying the most efficient oxidation process to achieve maximum removal of a target pollutant compound forms the subject of much research. There exists a need to develop rapid screening tools to support research in this area. In this work we report on the development of a quantitative assay as a means for identifying catalysts capable of decolourising methylene blue through the generation of oxidising species from hydrogen peroxide. Here, a previously described methylene blue test strip method was repurposed as a quantitative, aqueous-based spectrophotometric assay. From amongst a selection of metal salts and metallophthalocyanine complexes, monitoring of the decolourisation of the cationic dye methylene blue (via Fenton-like and non-Fenton oxidation reactions) by the assay identified the following to be suitable oxidation catalysts: CuSO
Sigma-Aldrich
29H,31H-Phthalocyanine, β-form, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
Manganese(II) phthalocyanine
Pricing and availability is not currently available.