Merck
Electronic properties of (Sb;Bi)
Scientific reports (2017-08-31)
Wasim J Mir, Alexandre Assouline, Clément Livache, Bertille Martinez, Nicolas Goubet, Xiang Zhen Xu, Gilles Patriarche, Sandrine Ithurria, Hervé Aubin, Emmanuel Lhuillier
ABSTRACT
We investigate the potential use of colloidal nanoplates of Sb
Sigma-Aldrich
Antimony(III) acetate, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, technical grade, 90%
Pricing and availability is not currently available.
Sigma-Aldrich
Bismuth(III) acetate, ≥99.99% trace metals basis
Pricing and availability is not currently available.