Merck
  • Survival role of locally produced acetylcholine in the bovine corpus luteum.
Survival role of locally produced acetylcholine in the bovine corpus luteum.
Biology of reproduction (2009-01-09)
M Omar Al-Zi'abi, Anom Bowolaksono, Kiyoshi Okuda
ABSTRACT
The present study was conducted to explore the source of acetylcholine (ACH) in the corpus luteum (CL) and to test our hypothesis of an antiapoptotic role of ACH in the bovine CL and, further, to investigate whether nerve growth factor (NGF), insulin-like growth factor 1 (IGF1), and transforming growth factor beta1 (TGFB1) influence the expression of choline acetyltransferase (CHAT), the biosynthetic enzyme of ACH, in cultured bovine luteal cells. Protein expression and immunolocalization of CHAT were carried out at different stages throughout the luteal phase and in cultured luteal and endothelial cells. ACH was measured in luteal tissue at the different luteal stages and in luteal cells cultured for 8 and 24 h. Cell viability and TUNEL assays were performed on cultured midluteal cells treated with or without tumor necrosis factor alpha (TNF)/interferon gamma (IFNG) in the presence of ACH and its muscarinic (atropine) and nicotinic (mecamylamine) receptor antagonists. The CL was devoid of cholinergic nerve fibers. CHAT immunostaining was evident in luteal, endothelial, and stromal cells in luteal tissue sections and in cultured luteal and endothelial cells. CHAT protein was expressed throughout the cycle without any significant changes. ACH concentration in luteal tissue was not changed during the luteal stages but increased over time and with increased cell numbers in luteal cell cultures. ACH increased cell viability and prevented cell death induced by TNF/IFNG. Atropine significantly attenuated ACH action, whereas mecamylamine had no effect. TNF/IFNG treatment downregulated CHAT expression, whereas NGF, IGF1, and TGFB1 upregulated CHAT expression, in cultured luteal cells. The overall findings strongly suggest a nonneural source and antiapoptotic role of ACH in the bovine CL. Locally produced ACH appears to be regulated by NGF, IGF1, and TGFB1.
Sigma-Aldrich
Acetylcholine chloride, suitable for cell culture
Sigma-Aldrich
Rabbit serum
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Atropine sulfate salt monohydrate, ≥97% (TLC), crystalline
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Sigma-Aldrich
Mecamylamine hydrochloride