Merck
  • The Transgenic Zebrafish Display Fluorescence Reflecting the Expressional Dynamics of Dihydrofolate Reductase.
The Transgenic Zebrafish Display Fluorescence Reflecting the Expressional Dynamics of Dihydrofolate Reductase.
Zebrafish (2017-03-30)
Wen-Ni Chang, Wan-Yu Chi, Tseng-Ting Kao, Jen-Ning Tsai, Wangta Liu, Shih-Shin Liang, Chien-Chih Chiu, Bing-Hung Chen, Tzu-Fun Fu
ABSTRACT
Dihydrofolate reductase (DHFR) reduces folic acid and recycles dihydrofolate generated during dTMP biosynthesis to tetrahydrofolate. DHFR is upregulated in rapidly proliferating cells and hence a favored target of antifolate drug against cancers, autoimmune diseases, and microbial infections. However, increased expression of dhfr contributed to the often emerging drug resistance and impeded the therapeutic efficacy of antifolate drugs. Therefore, comprehensive knowledge on the expressional control of dhfr becomes crucial. We generated two zebrafish transgenic lines, Tg(zdhfr
Sigma-Aldrich
Poly-L-lysine hydrobromide, mol wt 1,000-5,000
Pricing and availability is not currently available.