Merck
  • Procyanidin dimers A1, A2, and B2 are absorbed without conjugation or methylation from the small intestine of rats.
Procyanidin dimers A1, A2, and B2 are absorbed without conjugation or methylation from the small intestine of rats.
The Journal of nutrition (2009-06-06)
Maaike M Appeldoorn, Jean-Paul Vincken, Harry Gruppen, Peter C H Hollman
ABSTRACT
Intervention studies with procyanidin (PC)-rich extracts and products such as cocoa and wine suggest protective effects of PC against cardiovascular diseases. However, there is no consensus on the absorption and metabolism of PC dimers. Interestingly, nothing is known about the absorption of A-type PC. In this study, the absorption and metabolism of purified PC dimers A1 [epicatechin-(2-O-7, 4-8)-catechin], A2 [epicatechin-(2-O-7, 4-8)-epicatechin], and B2 [epicatechin-(4-8)-epicatechin], A-type trimers, a mixture of A1, B2, and a tetrameric A-type, and monomeric epicatechin were compared by in situ perfusion of the small intestine of rats for 0-30 min. The rats had their bile duct, portal vein, and small intestine cannulated. Unmodified and methylated metabolites were distinguished from their conjugates by differential beta-glucuronidase treatment. A1 and A2 dimers were absorbed from the small intestine of rats and they were better absorbed than dimer B2. Absorption of the A-type dimers was only 5-10% of that of monomeric epicatechin. Dimers were not conjugated or methylated in contrast to epicatechin, which was partly methylated and 100% conjugated. A-type trimers were not absorbed. Furthermore, the presence of tetrameric PC enhanced the absorption of B2 but not that of A1. Epicatechin, methylated epicatechin, and their conjugates were not found as metabolites of the PC tested. In conclusion, dimers A1, A2, and B2 are slightly absorbed but are not conjugated or methylated, thus conserving their biological activity after absorption. Because PC contents of foods are relatively high, dimers may contribute to systemic effects of PC.
Sigma-Aldrich
β-Glucuronidase from Helix pomatia, Type H-5, lyophilized powder, ≥400,000 units/g solid
Pricing and availability is not currently available.
Supelco
Procyanidin B2, analytical standard
Pricing and availability is not currently available.