Merck
  • Intravascular ultrasound-derived morphological predictors of myocardial ischemia assessed by stress myocardial perfusion computed tomography.
Intravascular ultrasound-derived morphological predictors of myocardial ischemia assessed by stress myocardial perfusion computed tomography.
Catheterization and cardiovascular interventions : official journal of the Society for Cardiac Angiography & Interventions (2016-08-28)
Soo-Jin Kang, Dong Hyun Yang, Hyun Jung Koo, Sung-Cheol Yun, June-Goo Lee, Joon-Won Kang, Tae-Hwan Lim, Gary S Mintz, Seong-Wook Park, Young-Hak Kim
ABSTRACT
Although ischemia-guided revascularization improves clinical outcomes, morphological determinants of clinically relevant myocardial ischemia have not been studied. To identify intravascular ultrasound (IVUS)-derived anatomical parameters for predicting myocardial perfusion defect and its extent. A total of 103 patients (88 stable and 15 unstable angina) with 153 lesions (angiographic diameter stenosis of 30-80%) underwent stress myocardial perfusion computed tomography (CT) and IVUS pre-procedure. The volume of CT perfusion defect and %CT perfusion defect in the target vessel territories were measured. The CT perfusion defect was seen in 76 (50%) lesions. The independent determinants for the presence of CT perfusion defect were IVUS-minimal lumen area (MLA) (adjusted OR = 0.56, 95% CI = 0.38-0.82), plaque burden (adjusted OR = 1.07, 95% CI = 1.02-1.11) and involvement of left main or left anterior descending artery (adjusted OR = 4.13, 95% CI = 1.75-9.78, all P < 0.05). The CT perfusion defect was predicted by IVUS-MLA <2.28mm IVUS-derived morphological parameters were useful to predict the presence of CT perfusion defect and the size of myocardial ischemia that were primarily determined by lesion severity and subtended myocardial territory. © 2016 Wiley Periodicals, Inc.
Product Description
Isosorbide dinitrate, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.