Merck
  • Non-targeted LC-MS based metabolomics analysis of the urinary steroidal profile.
Non-targeted LC-MS based metabolomics analysis of the urinary steroidal profile.
Analytica chimica acta (2017-03-30)
Amelia Palermo, Francesco Botrè, Xavier de la Torre, Nicola Zamboni
ABSTRACT
The urinary steroidal fraction has been extensively explored as non-invasive alternative to monitor pathological conditions as well as to unveil the illicit intake of pseudo-endogenous anabolic steroids in sport. However, the majority of previous approaches involved the a priori selection of potentially relevant target analytes. Here we describe the non-targeted analysis of the urinary steroidal profiles. The workflow includes minimal sample pretreatment and normalization according to the specific gravity of urine, a 20 min reverse phase ultra-performance liquid chromatographic separation hyphenated to electrospray time-of-flight mass spectrometry. As initial validation, we analyzed a set of quality control urines spiked with glucurono- and sulfo-conjugated steroids at physiological ranges. We then applied the method for the analysis of samples collected after single transdermal administration of testosterone in hypogonadal men. The method allowed profiling of approximately three thousand metabolic features, including steroids of clinical and forensic relevance. It successfully identified metabolic pathways mostly responsible for groups clustering even in the context of high inter-individual variability and allowed the detection of currently unknown metabolic features correlating with testosterone administration. These outcomes set the stage for future studies aimed at implementing currently monitored urinary steroidal markers both in clinical and forensic analysis.