Merck
NKG2D
Scientific reports (2017-04-30)
Di Yang, Zhiqiang Tian, Mengjie Zhang, Weibing Yang, Jun Tang, Yuzhang Wu, Bing Ni
ABSTRACT
Systemic lupus erythematosus (SLE) features a decreased pool of CD4
Sigma-Aldrich
Mouse Granzyme B ELISA Kit, for serum, plasma and cell culture supernatant
Pricing and availability is not currently available.