Merck
RBM19 is essential for preimplantation development in the mouse.
BMC developmental biology (2008-12-18)
Jian Zhang, Amber J Tomasini, Alan N Mayer
ABSTRACT
RNA-binding motif protein 19 (RBM19, NCBI Accession # NP_083038) is a conserved nucleolar protein containing 6 conserved RNA recognition motifs. Its biochemical function is to process rRNA for ribosome biogenesis, and it has been shown to play a role in digestive organ development in zebrafish. Here we analyzed the role of RBM19 during mouse embryonic development by generating mice containing a mutation in the Rbm19 locus via gene-trap insertion. Homozygous mutant embryos failed to develop beyond the morula stage, showing defective nucleologenesis, activation of apoptosis, and upregulation of P53 target genes. A unique feature of RBM19 is its localization to the cytoplasm in morula stage-embryos, whereas most other nucleolar proteins are localized to the nucleolar precursor body (NPB). The nucleoli in the Rbm19 mutant embryos remain immature, yet they can carry out rRNA synthesis. The timing of developmental arrest occurs after expression of the inner cell mass markers OCT3/4 and NANOG, but prior to the specification of trophectoderm as reflected by CDX2 expression. The data indicate that RBM19 is essential for preimplantation development, highlighting the importance of de novo nucleologenesis during this critical developmental stage.
Product Description
Roche
Proteinase K, recombinant, PCR Grade, Solution from Pichia pastoris
Pricing and availability is not currently available.
Sigma-Aldrich
Saponin from Quillaja sp., Sapogenin content 20-35 %
Pricing and availability is not currently available.