Merck
  • Functional inhibition of acid sphingomyelinase by Fluphenazine triggers hypoxia-specific tumor cell death.
Functional inhibition of acid sphingomyelinase by Fluphenazine triggers hypoxia-specific tumor cell death.
Cell death & disease (2017-03-31)
Saskia Klutzny, Ralf Lesche, Matthias Keck, Stefan Kaulfuss, Andreas Schlicker, Sven Christian, Carolyn Sperl, Roland Neuhaus, Jeffrey Mowat, Michael Steckel, Björn Riefke, Stefan Prechtl, Karsten Parczyk, Patrick Steigemann
ABSTRACT
Owing to lagging or insufficient neo-angiogenesis, hypoxia is a feature of most solid tumors. Hypoxic tumor regions contribute to resistance against antiproliferative chemotherapeutics, radiotherapy and immunotherapy. Targeting cells in hypoxic tumor areas is therefore an important strategy for cancer treatment. Most approaches for targeting hypoxic cells focus on the inhibition of hypoxia adaption pathways but only a limited number of compounds with the potential to specifically target hypoxic tumor regions have been identified. By using tumor spheroids in hypoxic conditions as screening system, we identified a set of compounds, including the phenothiazine antipsychotic Fluphenazine, as hits with novel mode of action. Fluphenazine functionally inhibits acid sphingomyelinase and causes cellular sphingomyelin accumulation, which induces cancer cell death specifically in hypoxic tumor spheroids. Moreover, we found that functional inhibition of acid sphingomyelinase leads to overactivation of hypoxia stress-response pathways and that hypoxia-specific cell death is mediated by the stress-responsive transcription factor ATF4. Taken together, the here presented data suggest a novel, yet unexplored mechanism in which induction of sphingolipid stress leads to the overactivation of hypoxia stress-response pathways and thereby promotes their pro-apoptotic tumor-suppressor functions to specifically kill cells in hypoxic tumor areas.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATF4
Pricing and availability is not currently available.
Sigma-Aldrich
N-Palmitoyl-D-sphingomyelin, ≥96.0% (TLC)
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Triton X-100, laboratory grade
Pricing and availability is not currently available.