- Gene Replacement and Fluorescent Labeling to Study the Functional Role of Exopolysaccharides in
Gene Replacement and Fluorescent Labeling to Study the Functional Role of Exopolysaccharides in
Frontiers in microbiology (2017-08-10)
Nuria Castro-Bravo, Claudio Hidalgo-Cantabrana, Miguel A Rodriguez-Carvajal, Patricia Ruas-Madiedo, Abelardo Margolles
PMID28790996
ABSTRACT
An extracellular layer of exopolysaccharides (EPS) covers the surface of some