Merck
  • Kinetics and mechanism of oxidation of the anti-tubercular prodrug isoniazid and its analog by iridium(iv) as models for biological redox systems.
Kinetics and mechanism of oxidation of the anti-tubercular prodrug isoniazid and its analog by iridium(iv) as models for biological redox systems.
Dalton transactions (Cambridge, England : 2003) (2017-06-18)
Jingran Dong, Yanli Ren, Sufang Sun, Jiao Yang, Chunxia Nan, Hongmei Shi, Jianzhong Xu, Jie Duan, Tiesheng Shi, Lars I Elding
ABSTRACT
A complex reaction mechanism of oxidation of the anti-tubercular prodrug isoniazid (isonicotinic hydrazide, INH) by [IrCl
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium hexachloroiridate(IV) hexahydrate, 99.9% trace metals basis
Pricing and availability is not currently available.