Merck
  • Metabolic Obesity, Adipose Inflammation and Elevated Breast Aromatase in Women with Normal Body Mass Index.
Metabolic Obesity, Adipose Inflammation and Elevated Breast Aromatase in Women with Normal Body Mass Index.
Cancer prevention research (Philadelphia, Pa.) (2017-03-09)
Neil M Iyengar, Kristy A Brown, Xi Kathy Zhou, Ayca Gucalp, Kotha Subbaramaiah, Dilip D Giri, Heba Zahid, Priya Bhardwaj, Nils K Wendel, Domenick J Falcone, Hanhan Wang, Samantha Williams, Michael Pollak, Monica Morrow, Clifford A Hudis, Andrew J Dannenberg
ABSTRACT
Obesity is associated with breast white adipose tissue (WAT) inflammation, elevated levels of the estrogen biosynthetic enzyme, aromatase, and systemic changes that have been linked to the pathogenesis of breast cancer. Here, we determined whether metabolic obesity, including changes in breast biology and systemic effects, occurs in a subset of women with normal body mass index (BMI). Breast WAT and fasting blood were collected from 72 women with normal BMI (<25 kg/m