Merck
  • Cytoplasmic overexpression of RNA-binding protein HuR is a marker of poor prognosis in meningioma, and HuR knockdown decreases meningioma cell growth and resistance to hypoxia.
Cytoplasmic overexpression of RNA-binding protein HuR is a marker of poor prognosis in meningioma, and HuR knockdown decreases meningioma cell growth and resistance to hypoxia.
The Journal of pathology (2017-05-12)
Guillaume Gauchotte, Sébastien Hergalant, Charlène Vigouroux, Jean-Matthieu Casse, Rémi Houlgatte, Tony Kaoma, Déborah Helle, Lydia Brochin, Fabien Rech, Matthieu Peyre, François Labrousse, Laurent Vallar, Jean-Louis Guéant, Jean-Michel Vignaud, Shyue-Fang Battaglia-Hsu
ABSTRACT
HuR regulates cytoplasmic mRNA stability and translatability, and the HuR expression level has been shown to correlate with poor disease outcome in several cancer types; however, the prognostic value and potential pro-oncogenic properties of HuR in meningioma remain unclear. Thus, in the present study, we analysed 85 meningioma tissue samples to establish the relationship between HuR expression, tumour cell proliferation, and/or patient survival. In addition, we examined the anti-proliferative effects of HuR knockdown in two meningioma cell lines (IOMM-Lee and Ben-Men-1) and conducted transcriptome-wide analyses (IOMM-Lee cells) to elucidate the molecular consequences of HuR knockdown. The results of the present study showed HuR cytoplasmic expression to correlate positively with tumour grade (p = 1.2 × 10
Sigma-Aldrich
MISSION® esiRNA, targeting human ELAVL1
Pricing and availability is not currently available.