Merck
Prominent Oncogenic Roles of EVI1 in Breast Carcinoma.
Cancer research (2017-02-18)
Hui Wang, Thorsten Schaefer, Martina Konantz, Martin Braun, Zsuzsanna Varga, Anna M Paczulla, Selina Reich, Francis Jacob, Sven Perner, Holger Moch, Tanja N Fehm, Lothar Kanz, Klaus Schulze-Osthoff, Claudia Lengerke
ABSTRACT
Overexpression of the EVI1 oncogene is associated typically with aggressive myeloid leukemia, but is also detectable in breast carcinoma where its contributions are unexplored. Analyzing a tissue microarray of 608 breast carcinoma patient specimens, we documented EVI1 overexpression in both estrogen receptor-positive (ER
Sigma-Aldrich
Aphidicolin, Ready Made Solution, from Nigrospora sphaerica
Pricing and availability is not currently available.