Merck
  • Polyphenol metabolism in differently colored cultivars of red currant (Ribes rubrum L.) through fruit ripening.
Polyphenol metabolism in differently colored cultivars of red currant (Ribes rubrum L.) through fruit ripening.
Planta (2017-03-21)
Zala Zorenc, Robert Veberic, Darinka Koron, Silvija Miosic, Olly Sanny Hutabarat, Heidi Halbwirth, Maja Mikulic-Petkovsek
ABSTRACT
Rare red currants colors caused by low anthocyanin content in the pink and a lack of anthocyanins in the white cultivar correlated with low ANS gene expression, enzyme activity, and increased sugar/acid ratios. Changes in the contents of polyphenols, sugars, and organic acids in berries of the three differently colored Ribes rubrum L. cultivars ('Jonkheer van Tets', 'Pink Champagne', and 'Zitavia') were determined by LC-MS and HPLC at 4 sampling times during the last month of fruit ripening. The activities of the main flavonoid enzymes, chalcone synthase/chalcone isomerase (CHS/CHI), flavanone 3-hydroxylase (FHT), and dihydroflavonol 4-reductase (DFR), and the expression of anthocyanidin synthase (ANS) were additionally measured. Despite many attempts, activities of flavonol synthase and glycosyltransferase did not show reliable results, the reason of which they could not be demonstrated in this study. The pink fruited cultivar 'Pink Champagne' showed generally lower enzyme activity than the red cultivar 'Jonkheer van Tets'. The white cultivar 'Zitavia' showed very low CHS/CHI activity and ANS expression and no FHT and DFR activities were detected. The DFR of R. rubrum L. clearly preferred dihydromyricetin as substrate although no 3',4',5'-hydroxylated anthocyanins were present. The anthocyanin content of the red cultivar slightly increased during the last three weeks of ripening and reached a maximum of 890 mg kg
Product Description
Supelco
Procyanidin B1, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Quercitrin hydrate, ≥78%
Pricing and availability is not currently available.