Merck
Modulation effect of acidulated human serum albumin on Cu
Journal of inorganic biochemistry (2017-04-04)
Baolong Xie, Fufeng Liu, Xiaoyan Dong, Yongjian Wang, Xiaoguang Margaret Liu, Yan Sun
ABSTRACT
Aggregation of amyloid β-proteins (Aβ) induced by Cu
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Pricing and availability is not currently available.
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
Diglycolic anhydride, technical grade, 90%
Pricing and availability is not currently available.