Merck
  • Cell cycle-dependent activity of the volume- and Ca2+-activated anion currents in Ehrlich lettre ascites cells.
Cell cycle-dependent activity of the volume- and Ca2+-activated anion currents in Ehrlich lettre ascites cells.
Journal of cellular physiology (2006-11-18)
Thomas Kjaer Klausen, Andreas Bergdahl, Charlotte Hougaard, Palle Christophersen, Stine F Pedersen, Else K Hoffmann
ABSTRACT
Recent evidence implicates the volume-regulated anion current (VRAC) and other anion currents in control or modulation of cell cycle progression; however, the precise involvement of anion channels in this process is unclear. Here, Cl- currents in Ehrlich Lettre Ascites (ELA) cells were monitored during cell cycle progression, under three conditions: (i) after osmotic swelling (i.e., VRAC), (ii) after an increase in the free intracellular Ca2+ concentration (i.e., the Ca2+-activated Cl- current, CaCC), and (iii) under steady-state isotonic conditions. The maximal swelling-activated VRAC current decreased in G1 and increased in early S phase, compared to that in G0. The isotonic steady-state current, which seems to be predominantly VRAC, also decreased in G1, and increased again in early S phase, to a level similar to that in G0. In contrast, the maximal CaCC current (500 nM free Ca2+ in the pipette), was unaltered from G0 to G1, but decreased in early S phase. A novel high-affinity anion channel inhibitor, the acidic di-aryl-urea NS3728, which inhibited both VRAC and CaCC, attenuated ELA cell growth, suggesting a possible mechanistic link between cell cycle progression and cell cycle-dependent changes in the capacity for conductive Cl- transport. It is suggested that in ELA cells, entrance into the S phase requires an increase in VRAC activity and/or an increased potential for regulatory volume decrease (RVD), and at the same time a decrease in CaCC magnitude.
Sigma-Aldrich
NS3728, ≥98% (HPLC)
Pricing and availability is not currently available.