Merck
  • AP2 α modulates cystic fibrosis transmembrane conductance regulator function in the human intestine.
AP2 α modulates cystic fibrosis transmembrane conductance regulator function in the human intestine.
Journal of cystic fibrosis : official journal of the European Cystic Fibrosis Society (2017-04-26)
Vandana Kumari, Shruti Desai, Nadia A Ameen
ABSTRACT
AP2 is a clathrin-based endocytic adaptor complex comprising α, β2, μ2 and σ2 subunits. μ2 regulates CFTR endocytosis. The α subunit interacts with CFTR in the intestine but its physiologic significance is unclear. CFTR short circuit current was measured in intestinal T84 cells following shRNA knock down of AP2α (AP2αKD). Clathrin-coated structures (CCS) were immunolabeled and quantified in AP2αKD intestinal Caco2BBe (C2BBe) cells. GST tagged human AP2α appendage domain was cloned and its interaction with CFTR determined by GST pull down assay. AP2αKD in T84 cells resulted in higher CFTR current (57%) compared to control, consistent with increased functional CFTR and delayed endocytosis. Depletion of AP2α reduced CCS in C2BBe cells. Pull down assays revealed an interaction between human AP2α appendage domain and CFTR. AP2 α interacts with and modulates CFTR function in the intestine by participating in clathrin assembly and recruitment of CFTR to CCS.
Sigma-Aldrich
MISSION® esiRNA, targeting human GTF3A
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Clathrin Heavy Chain Mouse mAb (X22), liquid, clone X22, Calbiochem®
Pricing and availability is not currently available.