Merck
  • Photobiomodulation of human dermal fibroblasts in vitro: decisive role of cell culture conditions and treatment protocols on experimental outcome.
Photobiomodulation of human dermal fibroblasts in vitro: decisive role of cell culture conditions and treatment protocols on experimental outcome.
Scientific reports (2017-06-07)
C Mignon, N E Uzunbajakava, B Raafs, N V Botchkareva, D J Tobin
ABSTRACT
Photobiomodulation-based (LLLT) therapies show tantalizing promise for treatment of skin diseases. Confidence in this approach is blighted however by lamentable inconsistency in published experimental designs, and so complicates interpretation. Here we interrogate the appropriateness of a range of previously-reported treatment parameters, including light wavelength, irradiance and radiant exposure, as well as cell culture conditions (e.g., serum concentration, cell confluency, medium refreshment, direct/indirect treatment, oxygen concentration, etc.), in primary cultures of normal human dermal fibroblasts exposed to visible and near infra-red (NIR) light. Apart from irradiance, all study parameters impacted significantly on fibroblast metabolic activity. Moreover, when cells were grown at atmospheric O
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - low glucose, With 1000 mg/L glucose, and sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture