Merck
  • Peptide dendrimer-conjugates of ketoprofen: Synthesis and ex vivo and in vivo evaluations of passive diffusion, sonophoresis and iontophoresis for skin delivery.
Peptide dendrimer-conjugates of ketoprofen: Synthesis and ex vivo and in vivo evaluations of passive diffusion, sonophoresis and iontophoresis for skin delivery.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2017-03-13)
Aswathi R Hegde, Prarthana V Rewatkar, Jyothsna Manikkath, Karnaker Tupally, Harendra S Parekh, Srinivas Mutalik
ABSTRACT
The aim of this study was to evaluate skin delivery of ketoprofen when covalently tethered to mildly cationic (2
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Triisopropylsilane, 98%
Sigma-Aldrich
Fmoc-Lys(Fmoc)-OH, ≥98.0% (HPLC)