Merck
  • Imaging and Detection of Carboxylesterase in Living Cells and Zebrafish Pretreated with Pesticides by a New Near-Infrared Fluorescence Off-On Probe.
Imaging and Detection of Carboxylesterase in Living Cells and Zebrafish Pretreated with Pesticides by a New Near-Infrared Fluorescence Off-On Probe.
Journal of agricultural and food chemistry (2017-05-06)
Dongyu Li, Zhao Li, Weihua Chen, Xingbin Yang
ABSTRACT
A new near-infrared fluorescence off-on probe was developed and applied to fluorescence imaging of carboxylesterase in living HepG-2 cells and zebrafish pretreated with pesticides (carbamate, organophosphorus, and pyrethroid). The probe was readily prepared by connecting (4-acetoxybenzyl)oxy as a quenching and recognizing moiety to a stable hemicyanine skeleton that can be formed via the decomposition of IR-780. The fluorescence off-on response of the probe to carboxylesterase is based on the enzyme-catalyzed spontaneous hydrolysis of the carboxylic ester bond, followed by a further fragmentation of the phenylmethyl unit and thereby the fluorophore release. Compared with the only existing near-infrared carboxylesterase probe, the proposed probe exhibits superior analytical performance, such as near-infrared fluorescence emission over 700 nm as well as high selectivity and sensitivity, with a detection limit of 4.5 × 10
Sigma-Aldrich
4-(Chloromethyl)phenyl acetate, 98%
Pricing and availability is not currently available.