Merck
  • Triglyceride-rich lipoprotein lipolysis products increase blood-brain barrier transfer coefficient and induce astrocyte lipid droplets and cell stress.
Triglyceride-rich lipoprotein lipolysis products increase blood-brain barrier transfer coefficient and induce astrocyte lipid droplets and cell stress.
American journal of physiology. Cell physiology (2017-01-13)
Linda L Lee, Hnin H Aung, Dennis W Wilson, Steven E Anderson, John C Rutledge, Jennifer M Rutkowsky
ABSTRACT
Elevation of blood triglycerides, primarily as triglyceride-rich lipoproteins (TGRL), has been linked to cerebrovascular inflammation, vascular dementia, and Alzheimer's disease (AD). Brain microvascular endothelial cells and astrocytes, two cell components of the neurovascular unit, participate in controlling blood-brain barrier (BBB) permeability and regulating neurovascular unit homeostasis. Our studies showed that infusion of high physiological concentrations of TGRL lipolysis products (TGRL + lipoprotein lipase) activate and injure brain endothelial cells and transiently increase the BBB transfer coefficient (Ki = permeability × surface area/volume) in vivo. However, little is known about how blood lipids affect astrocyte lipid accumulation and inflammation. To address this, we first demonstrated TGRL lipolysis products increased lipid droplet formation in cultured normal human astrocytes. We then evaluated the transcriptional pathways activated in astrocytes by TGRL lipolysis products and found upregulated stress and inflammatory-related genes including activating transcription factor 3 (ATF3), macrophage inflammatory protein-3α (MIP-3α), growth differentiation factor-15 (GDF15), and prostaglandin-endoperoxide synthase 2 (COX2). TGRL lipolysis products also activated the JNK/cJUN/ATF3 pathway, induced endoplasmic reticulum stress protein C/EBP homologous protein (CHOP), and the NF-κB pathway, while increasing secretion of MIP-3α, GDF15, and IL-8. Thus our results demonstrate TGRL lipolysis products increase the BBB transfer coefficient (Ki), induce astrocyte lipid droplet formation, activate cell stress pathways, and induce secretion of inflammatory cytokines. Our observations are consistent with evidence for lipid-induced neurovascular injury and inflammation, and we, therefore, speculate that lipid-induced astrocyte injury could play a role in cognitive decline.
Sigma-Aldrich
Free Glycerol Reagent, used for quantitative enzymatic determination of glycerol
Pricing and availability is not currently available.
Sigma-Aldrich
Glycerol Standard Solution, 2.5 mg/ml equivalent triolein concentration
Pricing and availability is not currently available.
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Lipoprotein Lipase from bovine milk, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Pricing and availability is not currently available.
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Triglyceride Reagent, used for quantitative enzymatic determination of triglyerides
Pricing and availability is not currently available.
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Pricing and availability is not currently available.
Supelco
Stearic acid, analytical standard
Pricing and availability is not currently available.
Sigma-Aldrich
Protease Inhibitor Cocktail Set I, A cocktail of five protease inhibitors that will inhibit a broad range of proteases and esterases. Supplied with a data sheet.
Pricing and availability is not currently available.