Merck
  • Synthesis, characterization and antifungal efficacy of C-coordinated O-carboxymethyl chitosan Cu(II) complexes.
Synthesis, characterization and antifungal efficacy of C-coordinated O-carboxymethyl chitosan Cu(II) complexes.
Carbohydrate polymers (2017-01-25)
Weixiang Liu, Yukun Qin, Song Liu, Ronge Xing, Huahua Yu, Xiaolin Chen, Kecheng Li, Pengcheng Li
ABSTRACT
A novel type of O-carboxymethyl chitosan Schiff bases (O-CSPX) was synthesized via a condensation reaction. After the coordination reaction of cupric ions, Cu(II) complexes (O-CSPX-Cu) were achieved. The theoretical structure of O-CSPX-Cu calculated by Gaussian 09 reveals that the copper ions underwent dsp
Sigma-Aldrich
4-Amino-2-methylbenzoic acid, 95%
Pricing and availability is not currently available.
Sigma-Aldrich
4-Amino-2-chlorobenzoic acid, 97%
Pricing and availability is not currently available.
Sigma-Aldrich
3-Amino-4-methylbenzoic acid, 99%
Pricing and availability is not currently available.