Merck
  • Loss of HSulf-1: The Missing Link between Autophagy and Lipid Droplets in Ovarian Cancer.
Loss of HSulf-1: The Missing Link between Autophagy and Lipid Droplets in Ovarian Cancer.
Scientific reports (2017-02-09)
Debarshi Roy, Susmita Mondal, Ashwani Khurana, Deok-Beom Jung, Robert Hoffmann, Xiaoping He, Eleftheria Kalogera, Thomas Dierks, Edward Hammond, Keith Dredge, Viji Shridhar
ABSTRACT
Defective autophagy and deranged metabolic pathways are common in cancer; pharmacologic targeting of these two pathways could provide a viable therapeutic option. However, how these pathways are regulated by limited availability of growth factors is still unknown. Our study shows that HSulf-1 (endosulfatase), a known tumor suppressor which attenuates heparin sulfate binding growth factor signaling, also regulates interplay between autophagy and lipogenesis. Silencing of HSulf-1 in OV202 and TOV2223 cells (ovarian cancer cell lines) resulted in increased lipid droplets (LDs), reduced autophagic vacuoles (AVs) and less LC3B puncta. In contrast, HSulf-1 proficient cells exhibit more AVs and reduced LDs. Increased LDs in HSulf-1 depleted cells was associated with increased ERK mediated cPLA2S505 phosphorylation. Conversely, HSulf-1 expression in SKOV3 cells reduced the number of LDs and increased the number of AVs compared to vector controls. Furthermore, pharmacological (AACOCF3) and ShRNA mediated downregulation of cPLA2 resulted in reduced LDs, and increased autophagy. Finally, in vivo experiment using OV202 Sh1 derived xenograft show that AACOCF3 treatment effectively attenuated tumor growth and LD biogenesis. Collectively, these results show a reciprocal regulation of autophagy and lipid biogenesis by HSulf-1 in ovarian cancer.
Sigma-Aldrich
Anti-phospho-c-PLA2 (pSer505) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
ApopTag Fluorescein In Situ Apoptosis Detection Kit, The ApopTag Fluorescein In Situ Apoptosis Detection Kit detects apoptotic cells in situ by the indirect TUNEL method, utilizing an anti-digoxigenin antibody that is conjugated to a Fluorescein reporter molecule.
Pricing and availability is not currently available.