Merck
  • Synthesis of Terephthalic Acid by p-Cymene Oxidation using Oxygen: Toward a More Sustainable Production of Bio-Polyethylene Terephthalate.
Synthesis of Terephthalic Acid by p-Cymene Oxidation using Oxygen: Toward a More Sustainable Production of Bio-Polyethylene Terephthalate.
ChemSusChem (2016-10-13)
Florentina Neaţu, Geanina Culică, Mihaela Florea, Vasile I Parvulescu, Fabrizio Cavani
ABSTRACT
The synthesis of terephthalic acid from biomass remains an unsolved challenge. In this study, we conducted the selective oxidation of p-cymene (synthesized from biodegradable terpenes, limonene, or eucalyptol) into terephthalic acid over a Mn-Fe mixed-oxide heterogeneous catalyst. The impact of various process parameters (oxidant, temperature, reaction time, catalyst amount, oxygen pressure) on the selectivity to terephthalic acid was evaluated, and some mechanistic aspects were elucidated. An unprecedented synthesis of biobased terephthalic acid (51 % yield) in the presence of O