Merck
  • Atrial natriuretic peptide regulates adipose tissue accumulation in adult atria.
Atrial natriuretic peptide regulates adipose tissue accumulation in adult atria.
Proceedings of the National Academy of Sciences of the United States of America (2017-01-18)
Nadine Suffee, Thomas Moore-Morris, Patrick Farahmand, Catherine Rücker-Martin, Gilles Dilanian, Magali Fradet, Daigo Sawaki, Geneviève Derumeaux, Pascal LePrince, Karine Clément, Isabelle Dugail, Michel Puceat, Stéphane N Hatem
ABSTRACT
The abundance of epicardial adipose tissue (EAT) is associated with atrial fibrillation (AF), the most frequent cardiac arrhythmia. However, both the origin and the factors involved in EAT expansion are unknown. Here, we found that adult human atrial epicardial cells were highly adipogenic through an epithelial-mesenchymal transition both in vitro and in vivo. In a genetic lineage tracing the WT1CreERT2+/-RosatdT+/- mouse model subjected to a high-fat diet, adipocytes of atrial EAT derived from a subset of epicardial progenitors. Atrial myocardium secretome induces the adipogenic differentiation of adult mesenchymal epicardium-derived cells by modulating the balance between mesenchymal Wingless-type Mouse Mammary Tumor Virus integration site family, member 10B (Wnt10b)/β-catenin and adipogenic ERK/MAPK signaling pathways. The adipogenic property of the atrial secretome was enhanced in AF patients. The atrial natriuretic peptide secreted by atrial myocytes is a major adipogenic factor operating at a low concentration by binding to its natriuretic peptide receptor A (NPRA) receptor and, in turn, by activating a cGMP-dependent pathway. Hence, our data indicate cross-talk between EAT expansion and mechanical function of the atrial myocardium.
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone VIM-13.2, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
8-Bromoguanosine 3′,5′-cyclic monophosphate sodium salt, ≥98% (HPLC), powder
Pricing and availability is not currently available.