Merck
  • Hydrogen bonding of water confined in mesoporous silica MCM-41 and SBA-15 studied by 1H solid-state NMR.
Hydrogen bonding of water confined in mesoporous silica MCM-41 and SBA-15 studied by 1H solid-state NMR.
Chemistry (Weinheim an der Bergstrasse, Germany) (2004-10-08)
Bob Grünberg, Thomas Emmler, Egbert Gedat, Ilja Shenderovich, Gerhard H Findenegg, Hans-Heinrich Limbach, Gerd Buntkowsky
ABSTRACT
The adsorption of water in two mesoporous silica materials with cylindrical pores of uniform diameter, MCM-41 and SBA-15, was studied by 1H MAS (MAS=magic angle spinning) and static solid-state NMR spectroscopy. All observed hydrogen atoms are either surface -SiOH groups or hydrogen-bonded water molecules. Unlike MCM-41, some strongly bound water molecules exist at the inner surfaces of SBA-15 that are assigned to surface defects. At higher filling levels, a further difference between MCM-41 and SBA-15 is observed. Water molecules in MCM-41 exhibit a bimodal line distribution of chemical shifts, with one peak at the position of inner-bulk water, and the second peak at the position of water molecules in fast exchange with surface -SiOH groups. In SBA-15, a single line is observed that shifts continuously as the pore filling is increased. This result is attributed to a different pore-filling mechanism for the two silica materials. In MCM-41, due to its small pore diameter (3.3 nm), pore filling by pore condensation (axial-pore-filling mode) occurs at a low relative pressure, corresponding roughly to a single adsorbed monolayer. For SBA-15, owing to its larger pore diameter (8 nm), a gradual increase in the thickness of the adsorbed layer (radial-pore-filling mode) prevails until pore condensation takes place at a higher level of pore filling.
Sigma-Aldrich
Silica, mesoporous SBA-15, <150 μm particle size, pore size 6 nm, amine functionalized
Pricing and availability is not currently available.
Sigma-Aldrich
Silica, mesoporous MCM-48, <15 μm particle size, pore size 3 nm, thiol functionalized
Pricing and availability is not currently available.