Merck
Distribution of local anesthetics between aqueous and liposome phases.
Journal of chromatography. A (2016-12-14)
Suvi-Katriina Ruokonen, Filip Duša, Antti H Rantamäki, Alexandra Robciuc, Paula Holma, Juha M Holopainen, Mohamed Abdel-Rehim, Susanne K Wiedmer
ABSTRACT
Liposomes were used as biomimetic models in capillary electrokinetic chromatography (EKC) for the determination of distribution constants (K
Sigma-Aldrich
Pentyl benzoate, AldrichCPR
Pricing and availability is not currently available.
Sigma-Aldrich
Propyl benzoate, 99%
Pricing and availability is not currently available.