Merck
  • Transgenic Expression of the Mitochondrial Chaperone TNFR-associated Protein 1 (TRAP1) Accelerates Prostate Cancer Development.
Transgenic Expression of the Mitochondrial Chaperone TNFR-associated Protein 1 (TRAP1) Accelerates Prostate Cancer Development.
The Journal of biological chemistry (2016-10-19)
Sofia Lisanti, David S Garlick, Kelly G Bryant, Michele Tavecchio, Gordon B Mills, Yiling Lu, Andrew V Kossenkov, Louise C Showe, Lucia R Languino, Dario C Altieri
ABSTRACT
Protein homeostasis, or proteostasis, is required for mitochondrial function, but its role in cancer is controversial. Here we show that transgenic mice expressing the mitochondrial chaperone TNFR-associated protein 1 (TRAP1) in the prostate develop epithelial hyperplasia and cellular atypia. When examined on a Pten+/- background, a common alteration in human prostate cancer, TRAP1 transgenic mice showed accelerated incidence of invasive prostatic adenocarcinoma, characterized by increased cell proliferation and reduced apoptosis, in situ Conversely, homozygous deletion of TRAP1 delays prostatic tumorigenesis in Pten+/- mice without affecting hyperplasia or prostatic intraepithelial neoplasia. Global profiling of Pten+/--TRAP1 transgenic mice by RNA sequencing and reverse phase protein array reveals modulation of oncogenic networks of cell proliferation, apoptosis, cell motility, and DNA damage. Mechanistically, reconstitution of Pten+/- prostatic epithelial cells with TRAP1 increases cell proliferation, reduces apoptosis, and promotes cell invasion without changes in mitochondrial bioenergetics. Therefore, TRAP1 is a driver of prostate cancer in vivo and an "actionable" therapeutic target.
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Roche
In Situ Cell Death Detection Kit, Fluorescein, sufficient for ≤50 tests, suitable for detection