Merck
  • Synthesis and cardiovascular protective effects of quercetin 7-O-sialic acid.
Synthesis and cardiovascular protective effects of quercetin 7-O-sialic acid.
Journal of cellular and molecular medicine (2016-08-12)
Hua Tian, Qingchao Liu, Shucun Qin, Chuanlong Zong, Ying Zhang, Shutong Yao, Nana Yang, Tao Guan, Shoudong Guo
ABSTRACT
Oxidative stress and inflammation play important roles in the pathogenesis of cardiovascular disease (CVD). Oxidative stress-induced desialylation is considered to be a primary step in atherogenic modification, and therefore, the attenuation of oxidative stress and/or inflammatory reactions may ameliorate CVD. In this study, quercetin 7-O-sialic acid (QA) was synthesized aiming to put together the cardiovascular protective effect of quercetin and the recently reported anti-oxidant and anti-atherosclerosis functions of N-acetylneuraminic acid. The biological efficacy of QA was evaluated in vitro in various cellular models. The results demonstrated that 50 μM QA could effectively protect human umbilical vein endothelial cells (HUVEC, EA.hy926) against hydrogen peroxide- or oxidized low-density lipoprotein-induced oxidative damage by reducing the production of reactive oxygen species. QA attenuated hydrogen peroxide-induced desialylation of HUVEC and lipoproteins. QA decreased lipopolysaccharide-induced secretion of tumour necrosis factor-α (TNF-α) and monocyte chemoattractant protein-1 (MCP-1), and it significantly reduced the expression of intercellular adhesion molecule-1, vascular cell adhesion molecule-1, TNF-α and MCP-1. Furthermore, QA effectively promoted cholesterol efflux from Raw 264.7 macrophages to apolipoprotein A-1 and high-density lipoprotein by up-regulating ATP-binding cassette transporter A1 and G1, respectively. Results indicated that the novel compound QA exhibited a better capacity than quercetin for anti-oxidation, anti-inflammation, cholesterol efflux promotion and biomolecule protection against desialylation and therefore could be a candidate compound for the prevention or treatment of CVD.
Sigma-Aldrich
Fluorescein-5-thiosemicarbazide, suitable for fluorescence, ~80% (HPCE)
Pricing and availability is not currently available.