Merck
  • Hypochlorite converts cysteinyl-dopamine into a cytotoxic product: A possible factor in Parkinson's Disease.
Hypochlorite converts cysteinyl-dopamine into a cytotoxic product: A possible factor in Parkinson's Disease.
Free radical biology & medicine (2016-11-05)
Nihar J Mehta, Karam Asmaro, David J Hermiz, Meredith M Njus, Ashraf H Saleh, Karen A Beningo, David Njus
ABSTRACT
The dopamine oxidation product cysteinyl-dopamine has attracted attention as a contributor to the death of dopaminergic neurons in Parkinson's disease. Treatment of cysteinyl-dopamine with hypochlorite yields an even more cytotoxic product. This product has potent redox-cycling activity and initiates production of superoxide in PC12 cells. Taurine, which scavenges hypochlorite, protects PC12 cells from cysteinyl-dopamine but not from the hypochlorite product, suggesting that the product, not cysteinyl-dopamine itself, is toxic. Furthermore, rotenone, which enhances expression of the hypochlorite-producing enzyme myeloperoxidase, increases the cytotoxicity of cysteinyl-dopamine but not of the hypochlorite product. This suggests that dopamine oxidation to cysteinyl-dopamine followed by hypochlorite-dependent conversion to a cytotoxic redox-cycling product leads to the generation of reactive oxygen species and oxidative stress and may contribute to the death of dopaminergic neurons.
Sigma-Aldrich
SIGMAFAST Protease Inhibitor Tablets, For General Use