Merck
  • Trans-splicing into highly abundant albumin transcripts for production of therapeutic proteins in vivo.
Trans-splicing into highly abundant albumin transcripts for production of therapeutic proteins in vivo.
Molecular therapy : the journal of the American Society of Gene Therapy (2008-12-11)
Jun Wang, S Gary Mansfield, Colette A Cote, Ping Du Jiang, Ke Weng, Marcelo J A Amar, Bryan H Brewer, Alan T Remaley, Gerard J McGarrity, Mariano A Garcia-Blanco, M Puttaraju
ABSTRACT
Spliceosome-mediated RNA trans-splicing has emerged as an exciting mode of RNA therapy. Here we describe a novel trans-splicing strategy, which targets highly abundant pre-mRNAs, to produce therapeutic proteins in vivo. First, we used a pre-trans-splicing molecule (PTM) that mediated trans-splicing of human apolipoprotein A-I (hapoA-I) into the highly abundant mouse albumin exon 1. Hydrodynamic tail vein injection of the hapoA-I PTM plasmid in mice followed by analysis of the chimeric transcripts and protein, confirmed accurate and efficient trans-splicing into albumin pre-mRNA and production of hapoA-I protein. The versatility of this approach was demonstrated by producing functional human papillomavirus type-16 E7 (HPV16-E7) single-chain antibody in C57BL/6 mice and functional factor VIII (FVIII) and phenotypic correction in hemophilia A mice. Altogether, these studies demonstrate that trans-splicing to highly abundant albumin transcripts can be used as a general platform to produce therapeutic proteins in vivo.
Sigma-Aldrich
In Vitro Toxicology Assay Kit, MTT based
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.