Merck
  • BIX-01294 induces autophagy-associated cell death via EHMT2/G9a dysfunction and intracellular reactive oxygen species production.
BIX-01294 induces autophagy-associated cell death via EHMT2/G9a dysfunction and intracellular reactive oxygen species production.
Autophagy (2013-12-11)
Yunha Kim, Yong-Sook Kim, Dong Eun Kim, Jee Suk Lee, Ji Hoon Song, Ha-Gyeong Kim, Dong-Hyung Cho, Seong-Yun Jeong, Dong-Hoon Jin, Se Jin Jang, Hyang-Sook Seol, Young-Ah Suh, Seung Jin Lee, Choung-Soo Kim, Jae-Young Koh, Jung Jin Hwang
ABSTRACT
We screened a chemical library in MCF-7 cells stably expressing green fluorescent protein (GFP)-conjugated microtubule-associated protein 1 light chain 3 (LC3) (GFP-LC3-MCF-7) using cell-based assay, and identified BIX-01294 (BIX), a selective inhibitor of euchromatic histone-lysine N-methyltransferase 2 (EHMT2), as a strong autophagy inducer. BIX enhanced formation of GFP-LC3 puncta, LC3-II, and free GFP, signifying autophagic activation. Inhibition of these phenomena with chloroquine and increasement in punctate dKeima ratio (550/438) signal indicated that BIX activated autophagic flux. BIX-induced cell death was suppressed by the autophagy inhibitor, 3-methyladenine, or siRNA against BECN1 (VPS30/ATG6), ATG5, and ATG7, but not by caspase inhibitors. Moreover, EHMT2 siRNA augmented GFP-LC3 puncta, LC3-II, free GFP, and cell death, implying that inhibition of EHMT2 caused autophagy-mediated cell death. Treatment with EHMT2 siRNA and BIX accumulated intracellular reactive oxygen species (ROS). BIX augmented mitochondrial superoxide via NADPH oxidase activation. In addition, BIX increased hydrogen peroxide and glutathione redox potential in both cytosol and mitochondria. Treatment with N-acetyl-L-cysteine (NAC) or diphenyleneiodonium chloride (DPI) decreased BIX-induced LC3-II, GFP-LC3 puncta, and cell death, indicating that ROS instigated autophagy-dependent cell death triggered by BIX. We observed that BIX potentiated autophagy-dependent and caspase-independent cell death in estrogen receptor (ESR)-negative SKBr3 and ESR-positive MCF-7 breast cancer cells, HCT116 colon cancer cells, and importantly, in primary human breast and colon cancer cells. Together, the results suggest that BIX induces autophagy-dependent cell death via EHMT2 dysfunction and intracellular ROS accumulation in breast and colon cancer cells, therefore EHMT2 inhibition can be an effective therapeutic strategy for cancer treatment.
Sigma-Aldrich
N-Acetyl-L-cysteine, BioReagent, suitable for cell culture
Sigma-Aldrich
Anti-Actin (20-33) antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-dimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-G9a (BAT8) Antibody, from rabbit, purified by affinity chromatography