Merck
  • c-Myc and ChREBP regulate glucose-mediated expression of the L-type pyruvate kinase gene in INS-1-derived 832/13 cells.
c-Myc and ChREBP regulate glucose-mediated expression of the L-type pyruvate kinase gene in INS-1-derived 832/13 cells.
American journal of physiology. Endocrinology and metabolism (2007-03-08)
J Jason Collier, Pili Zhang, Kim B Pedersen, Susan J Burke, John W Haycock, Donald K Scott
ABSTRACT
Increased glucose flux generates metabolic signals that control transcriptional programs through poorly understood mechanisms. Previously, we demonstrated a necessity in hepatocytes for c-Myc in the regulation of a prototypical glucose-responsive gene, L-type pyruvate kinase (L-PK) (Collier JJ, Doan TT, Daniels MC, Schurr JR, Kolls JK, Scott DK. J Biol Chem 278: 6588-6595, 2003). Pancreatic beta-cells have many features in common with hepatocytes with respect to glucose-regulated gene expression, and in the present study we determined whether c-Myc was required for the L-PK glucose response in insulin-secreting (INS-1)-derived 832/13 cells. Glucose increased c-Myc abundance and association with its heterodimer partner, Max. Manipulations that prevented the formation of a functional c-Myc/Max heterodimer reduced the expression of the L-PK gene. In addition, glucose augmented the binding of carbohydrate response element binding protein (ChREBP), c-Myc, and Max to the promoter of the L-PK gene in situ. The transactivation of ChREBP, but not of c-Myc, was dependent on high glucose concentrations in the contexts of either the L-PK promoter or a heterologous promoter. The glucose-mediated transactivation of ChREBP was independent of mutations that alter phosphorylation sites thought to regulate the cellular location of ChREBP. We conclude that maximal glucose-induced expression of the L-PK gene in INS-1-derived 832/13 cells involves increased c-Myc abundance, recruitment of c-Myc, Max, and ChREBP to the promoter, and a glucose-stimulated increase in ChREBP transactivation.
Sigma-Aldrich
Chromatin Immunoprecipitation (ChIP) Assay Kit, Contains all necessary reagents to perform 22 individual chromatin immunoprecipitation (ChIP) reactions using inexpensive protein A agarose beads.
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
High Salt Immune Complex Wash Buffer, For use in the Chromatin Immunoprecipitation Assay Kits.