Merck
  • Mutations conferring zanamivir resistance in human influenza virus N2 neuraminidases compromise virus fitness and are not stably maintained in vitro.
Mutations conferring zanamivir resistance in human influenza virus N2 neuraminidases compromise virus fitness and are not stably maintained in vitro.
The Journal of antimicrobial chemotherapy (2006-08-08)
Thomas Zürcher, Phillip J Yates, Janet Daly, Anjali Sahasrabudhe, Matthew Walters, Laura Dash, Margaret Tisdale, Jennifer L McKimm-Breschkin
ABSTRACT
Viruses resistant to zanamivir have been generated in vitro, but no resistant virus has yet been isolated from a zanamivir-treated immunocompetent patient. In contrast most resistant viruses isolated from oseltamivir-treated patients correspond to those selected in vitro. However, despite mutations being in conserved residues in the neuraminidase (NA) they do not confer resistance in all NA subtypes. We have used reverse genetics and the recombinant baculovirus expression system for investigating reasons for the lack of isolation of zanamivir-resistant H3N2 viruses and for further exploring subtype-specific oseltamivir resistance. H3N2 viruses generated by reverse genetics with H274Y, R292K E119V and E119D mutations were rescued. Those with E119G, E119A or R152K mutations could only be rescued in the presence of exogenous NA and after passage in the absence of exogenous NA only isolates that had reverted to the wild-type NA or, surprisingly, E119G/A to E119V NA were isolated. Mutations conferring zanamivir resistance significantly affected enzyme activity, virus replication or NA thermal stability. E119V viruses were stable and grew to similar titres as wild-type virus, consistent with their isolation from oseltamivir-treated patients. Mutations conferring oseltamivir resistance in N1 (H274Y) and B (R152K) NAs also conferred resistance in recombinant G70C N9 NA expressed in insect cells. These data suggest that zanamivir-resistant H3N2 viruses may not readily arise in vivo due to their poor viability. The G70C N9 NA may also provide a useful model for understanding the structural basis of subtype-specific drug resistance.
Sigma-Aldrich
IgG from sheep serum, technical grade, ≥80% (SDS-PAGE), buffered aqueous solution