Merck
  • Epithelial to mesenchymal transition in human breast cancer can provide a nonmalignant stroma.
Epithelial to mesenchymal transition in human breast cancer can provide a nonmalignant stroma.
The American journal of pathology (2003-01-28)
Ole William Petersen, Helga Lind Nielsen, Thorarinn Gudjonsson, René Villadsen, Fritz Rank, Erik Niebuhr, Mina J Bissell, Lone Rønnov-Jessen
ABSTRACT
A breast carcinoma biopsy showed cytochemical evidence of epithelial mesenchymal transition and an alpha-smooth muscle actin-positive stromal reaction. To study the lineage, and the nature of the cells in the stromal reaction, we derived a novel cell line, HBFL-1, from the explanted biopsy. HBFL-1 cells are immortal and exhibit a shared non-random X-chromosome inactivation pattern with the epithelial tumor of origin. Yet they closely resemble normal, finite-life-span fibroblasts by morphology, lack of tumor formation in nude mice, marker expression profile, protein pattern using two-dimensional gel electrophoresis and the ability to undergo myofibroblast conversion. HBFL-1 interacts reciprocally with tumor cells in collagen gel to induce activation of MMP2, leading to tumor-like behavior of epithelial colonies. In vivo, HBFL-1 cells resembled normal-derived myofibroblasts and conferred a significant 3.5- to 7-fold increase in MCF-7 tumor size in nude mice. However, that they were indeed not normal fibroblasts was revealed by residual keratin expression and formation of epithelial microfoci in a reconstituted basement membrane and in nude mice. We conclude that breast cancer can generate its own nonmalignant stroma and that one function for this is that of a reciprocal interaction with epithelial tumor cells to facilitate tumor growth.
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid
Sigma-Aldrich
Monoclonal Anti-Fibroblast Surface Protein antibody produced in mouse, clone 1B10, ascites fluid