Merck
  • A model for regulation by SynGAP-α1 of binding of synaptic proteins to PDZ-domain 'Slots' in the postsynaptic density.
A model for regulation by SynGAP-α1 of binding of synaptic proteins to PDZ-domain 'Slots' in the postsynaptic density.
eLife (2016-10-21)
Ward G Walkup, Tara L Mastro, Leslie T Schenker, Jost Vielmetter, Rebecca Hu, Ariella Iancu, Meera Reghunathan, Barry Dylan Bannon, Mary B Kennedy
ABSTRACT
SynGAP is a Ras/Rap GTPase-activating protein (GAP) that is a major constituent of postsynaptic densities (PSDs) from mammalian forebrain. Its α1 isoform binds to all three PDZ (PSD-95, Discs-large, ZO-1) domains of PSD-95, the principal PSD scaffold, and can occupy as many as 15% of these PDZ domains. We present evidence that synGAP-α1 regulates the composition of the PSD by restricting binding to the PDZ domains of PSD-95. We show that phosphorylation by Ca2+/calmodulin-dependent protein kinase II (CaMKII) and Polo-like kinase-2 (PLK2) decreases its affinity for the PDZ domains by several fold, which would free PDZ domains for occupancy by other proteins. Finally, we show that three critical postsynaptic signaling proteins that bind to the PDZ domains of PSD-95 are present in higher concentration in PSDs isolated from mice with a heterozygous deletion of synGAP.
Sigma-Aldrich
Anti-Stargazin Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Rosetta 2(DE3) Competent Cells - Novagen, Novagen′s Rosetta 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. coli.
Pricing and availability is not currently available.
Sigma-Aldrich
Cdk5/p35 Protein, active, 10 µg, Active complex of recombinant full length human Cdk5 containing & N-terminal His6-tag & recombinant full-length human p35 containing an N-terminal GST-tag, for use in Kinase Assays.
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Neuroligin 1 antibody produced in mouse, clone S97A-31, purified immunoglobulin
Pricing and availability is not currently available.