Merck
  • A mitogen-activated protein kinase signals to programmed cell death induced by self-incompatibility in Papaver pollen.
A mitogen-activated protein kinase signals to programmed cell death induced by self-incompatibility in Papaver pollen.
Plant physiology (2007-07-31)
Shutian Li, Jozef Samaj, Vernonica E Franklin-Tong
ABSTRACT
Self-incompatibility (SI) in higher plants is an important mechanism to prevent inbreeding and involves specific rejection of incompatible ("self") pollen. In field poppy (Papaver rhoeas), S proteins encoded by the stigma component of the S-locus interact with incompatible pollen, resulting in cessation of tip growth. This "self" interaction triggers a Ca(2+)-dependent signaling network, involving programmed cell death (PCD). We previously identified p56, a mitogen-activated protein kinase (MAPK) that is activated during the SI response in incompatible pollen. Here, we show that p56 cross-reacts with AtMPK3, but not with AtMPK4 or salicylic acid-induced protein kinase antibodies. We provide good evidence that a MAPK is involved in initiation of SI-induced PCD in incompatible pollen. SI rapidly reduces pollen viability and the MAPK cascade inhibitor U0126, which prevents the SI-induced activation of p56 in incompatible pollen, "rescues" incompatible pollen, while its negative analog, U0124, does not. This strongly implicates the involvement of a MAPK in SI-mediated loss of pollen viability and cell death. SI also stimulates caspase-3-like (DEVDase) activity and later DNA fragmentation. Both these markers of PCD are significantly reduced by pretreatment with U0126, implicating the involvement of a MAPK in signaling during early PCD. As p56 appears to be the only MAPK activated by SI, our studies imply that p56 could be the MAPK involved in mediating SI-induced PCD.
Sigma-Aldrich
Anti-AtMPK4 antibody produced in rabbit, affinity isolated antibody, liquid