Merck
  • Heterogeneous architecture of vertebrate kinetochores revealed by three-dimensional superresolution fluorescence microscopy.
Heterogeneous architecture of vertebrate kinetochores revealed by three-dimensional superresolution fluorescence microscopy.
Molecular biology of the cell (2016-11-05)
David J Wynne, Hironori Funabiki
ABSTRACT
The kinetochore is often depicted as having a disk-like architecture in which the outer layer of proteins, which engage microtubules and control checkpoint signaling, are built on a static inner layer directly linked to CENP-A chromatin. Here, applying three-dimensional (3D) structural illumination microscopy (SIM) and stochastic optical reconstruction microscopy (STORM) to Xenopus egg extracts and tissue culture cells, we report various distribution patterns of inner and outer kinetochore proteins. In egg extracts, a configuration in which outer kinetochore proteins surround the periphery of CENP-A chromatin is common, forming an ∼200-nm ring-like organization that may engage a bundle of microtubule ends. Similar rings are observed in Xenopus tissue culture cells at a lower frequency but are enriched in conditions in which the spindle is disorganized. Although rings are rare in human cells, the distribution of both inner and outer kinetochore proteins elongates in the absence of microtubule attachment in a manner dependent on Aurora B. We propose a model in which the 3D organization of both the outer and inner kinetochore regions respond to the progression from lateral to end-on microtubule attachments by coalescing into a tight disk from less uniform distributions early in prometaphase.
Sigma-Aldrich
Glucose Oxidase from Aspergillus niger, Type VII, lyophilized powder, ≥100,000 units/g solid (without added oxygen)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-BubR1 Antibody, clone 8G1, clone 8G1, Chemicon®, from mouse
Pricing and availability is not currently available.