Merck
  • Activation of glycogen synthase kinase-3 inhibits long-term potentiation with synapse-associated impairments.
Activation of glycogen synthase kinase-3 inhibits long-term potentiation with synapse-associated impairments.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2007-11-09)
Ling-Qiang Zhu, Shao-Hui Wang, Dan Liu, Yang-Yang Yin, Qing Tian, Xiao-Chuan Wang, Qun Wang, Jian-Guo Chen, Jian-Zhi Wang
ABSTRACT
Activation of glycogen synthase kinase-3 (GSK-3) can cause memory deficits as seen in Alzheimer's disease, the most common age-associated dementia, but the mechanism is not understood. Here, we found that activation of GSK-3 by wortmannin or transient overexpression of wild-type GSK-3beta could suppress the induction of long-term potentiation (LTP) in rat hippocampus, whereas simultaneous inhibition of GSK-3 by lithium or SB216763 or transient expression of a dominant-negative GSK-3beta mutant (dnGSK-3beta) preserved the LTP. After high-frequency stimulation (HFS), the presynaptic release of glutamate and the expression/clustering of synapsin I, a synaptic vesicle protein playing an important role in neurotransmitter release, decreased markedly after upregulation of GSK-3. In vitro studies further demonstrated that GSK-3 inhibited the expression of SynI independent of HFS. In postsynaptic level, the expression of PSD93 and NR2A/B proteins decreased significantly when GSK-3 was activated. The LTP-associated synapse impairments including less presynaptic active zone, thinner postsynaptic density, and broader synaptic cleft were also prominent in the hippocampal slices after HFS with activation of GSK-3. These synaptic impairments were attenuated when GSK-3 was simultaneously inhibited by LiCl or SB216763 or transient expression of dnGSK-3. We conclude that upregulation of GSK-3 impairs the synaptic plasticity both functionally and structurally, which may underlie the GSK-3-involved memory deficits.
Sigma-Aldrich
Wortmannin, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Pricing and availability is not currently available.