Merck
  • Dynamics of the mouse brain cortical synaptic proteome during postnatal brain development.
Dynamics of the mouse brain cortical synaptic proteome during postnatal brain development.
Scientific reports (2016-10-18)
Miguel A Gonzalez-Lozano, Patricia Klemmer, Titia Gebuis, Chopie Hassan, Pim van Nierop, Ronald E van Kesteren, August B Smit, Ka Wan Li
ABSTRACT
Development of the brain involves the formation and maturation of numerous synapses. This process requires prominent changes of the synaptic proteome and potentially involves thousands of different proteins at every synapse. To date the proteome analysis of synapse development has been studied sparsely. Here, we analyzed the cortical synaptic membrane proteome of juvenile postnatal days 9 (P9), P15, P21, P27, adolescent (P35) and different adult ages P70, P140 and P280 of C57Bl6/J mice. Using a quantitative proteomics workflow we quantified 1560 proteins of which 696 showed statistically significant differences over time. Synaptic proteins generally showed increased levels during maturation, whereas proteins involved in protein synthesis generally decreased in abundance. In several cases, proteins from a single functional molecular entity, e.g., subunits of the NMDA receptor, showed differences in their temporal regulation, which may reflect specific synaptic development features of connectivity, strength and plasticity. SNARE proteins, Snap 29/47 and Stx 7/8/12, showed higher expression in immature animals. Finally, we evaluated the function of Cxadr that showed high expression levels at P9 and a fast decline in expression during neuronal development. Knock down of the expression of Cxadr in cultured primary mouse neurons revealed a significant decrease in synapse density.
Sigma-Aldrich
MISSION® TRC2 pLKO.5-puro TurboGFP shRNA Control Plasmid DNA, shRNA sequence targeting tGFP
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Growth Associated Protein-43 antibody produced in mouse, clone GAP-7B10, ascites fluid
Pricing and availability is not currently available.