Merck
  • Targeted deletion of ROCK1 protects the heart against pressure overload by inhibiting reactive fibrosis.
Targeted deletion of ROCK1 protects the heart against pressure overload by inhibiting reactive fibrosis.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2006-05-06)
Ying-Min Zhang, Jacqueline Bo, George E Taffet, Jiang Chang, Jianjian Shi, Anilkumar K Reddy, Lloyd H Michael, Michael D Schneider, Mark L Entman, Robert J Schwartz, Lei Wei
ABSTRACT
Ventricular myocyte hypertrophy is an important compensatory growth response to pressure overload. However, pathophysiological cardiac hypertrophy is accompanied by reactive fibrosis and remodeling. The Rho kinase family, consisting of ROCK1 and ROCK2, has been implicated in cardiac hypertrophy and ventricular remodeling. However, these previous studies relied heavily on pharmacological inhibitors,and not on gene deletion. Here we used ROCK1knockout (ROCK1-/-) mice to investigate role of ROCK1 in the development of ventricular remodeling induced by transverse aortic banding. We observed that ROCK1 deletion did not impair compensatory hypertrophic response induced by pressure overload. However, ROCK1-/- mice exhibited reduced perivascular and interstitial fibrosis, which was observed at 3 wk but not at 1 wk after the banding. The reduced fibrosis in the myocardium of ROCK1-/- mice was closely associated with reduced expression of a variety of extracellular matrix (ECM) proteins and fibrogenic cytokines such as TGFbeta2 and connective tissue growth factor. This inhibitory effect of ROCK1 deletion on pathophysiological induction of fibrogenic cytokines was further confirmed in the myocardium of transgenic mice with cardiomyocyte-specific overexpression of Gq. Thus, these results indicate that ROCK1 contributes to the development of cardiac fibrosis and induction of fibrogenic cytokines in cardiomyocytes in response to pathological stimuli.
Sigma-Aldrich
ROCK1 (17-535), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Pricing and availability is not currently available.